Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Distemper Virus Antibody (5-4), Alexa Fluor™ 405, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NB1102604AF405
Description
Distemper Virus Monoclonal specifically detects Distemper Virus in Virus samples. It is validated for ELISA, Sandwich ELISA.Specifications
Distemper Virus | |
Monoclonal | |
Alexa Fluor 405 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
Hybridoma clone has been derived from hybridization of Sp2/0 myeloma cells with spleen cells of Balb/c mice immunized with Canine distemper virus (CDV). | |
0.2 mL | |
Primary | |
Virus | |
Purified |
ELISA | |
43224 | |
ELISA, Sandwich ELISA | |
Mouse | |
Protein A purified | |
RUO | |
Specific for surface envelope antigen of CDV | |
Store at 4C in the dark. | |
IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction