Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Distemper Virus Antibody (5-4), Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NB1102604
Description
Distemper Virus Monoclonal specifically detects Distemper Virus in Virus samples. It is validated for ELISA, Immunoassay, Sandwich ELISA, Lateral Flow Assay.Specifications
| Distemper Virus | |
| Monoclonal | |
| Unconjugated | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Virus | |
| Purified |
| ELISA | |
| 43224 | |
| Sandwich ELISA 1:100-1:2000 | |
| Hybridoma clone has been derived from hybridization of Sp2/0 myeloma cells with spleen cells of Balb/c mice immunized with Canine distemper virus (CDV). | |
| 0.2 mg | |
| Virology Bacteria and Parasites | |
| Specific for surface envelope antigen of CDV | |
| Store at 4C. Do not freeze. | |
| IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction