Learn More
Description
- Produced through a distillation process which removes dissolved minerals, salts and many contaminants
- Useful in a wide range of laboratory applications
- Certificate of Analysis available
- Low ionic content helps minimize analytical interference supporting reliable results
- Induction sealed to ensure product integrity and decrease leakage during shipment
Specifications
Specifications
| Chemical Name or Material | Distilled Water |
| CAS | 7732-18-5 |
| Molecular Weight (g/mol) | 18 |
| Percent Purity | High Purity |
| Grade | Not Available |
| Quantity | 4 L |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
