Learn More
Synthware™ Vacuum Distillation Apparatus
GREENER_CHOICESDP
Catalog No. 31501041
Click to view available options
Standard Taper Joint:
10/18, 14/20
10/18, 19/22
14/20, 14/20
10/18 joint on side tube to accommodate a 25 mm immersion thermometer.
Specifications
For Use With (Application) Organic Synthesis
Type Vacuum Distillation Apparatus
Material Borosilicate Glass
For Use With (Equipment) 25 mm immersion thermometer
Quantity 1 Ea.
Standard Taper Joint 10/18, 19/22
Joint Type Ground Glass
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.