Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Disuccinimidyl suberate, 97%

Catalog No. aah51771me
Click to view available options
Quantity:
500 mg
1 g

CAS: 68528-80-3 | C16H20N2O8 | 368.342 g/mol

Disuccinimidyl suberate is a homobifunctional amine-reactive crosslinker. It is usually coupled to molecules containing primary amines via amide bonds buffered at pH 7.5 (6.5-8.5). It incorporates an 8-atom spacer arm.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Disuccinimidyl suberate is a homobifunctional amine-reactive crosslinker. It is usually coupled to molecules containing primary amines via amide bonds buffered at pH 7.5 (6.5-8.5). It incorporates an 8-atom spacer arm.

Solubility
Insoluble in water. Soluble in chloroform: 50 mg/mL.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 68528-80-3
Molecular Formula C16H20N2O8
Molecular Weight (g/mol) 368.342
MDL Number MFCD00049059
InChI Key ZWIBGKZDAWNIFC-UHFFFAOYSA-N
Synonym disuccinimidyl suberate, bis 2,5-dioxopyrrolidin-1-yl octanedioate, suberic acid bis n-hydroxysuccinimide ester, di n-succinimidyl suberate, bicl100, 2,5-pyrrolidinedione, 1,1'-1,8-dioxo-1,8-octanediyl bis oxy bis, nhs-sa, disuccinimidylsuberate, dsis, n-hydroxysuccinimide suberic acid ester
PubChem CID 100658
IUPAC Name bis(2,5-dioxopyrrolidin-1-yl) octanedioate
SMILES C1CC(=O)N(C1=O)OC(=O)CCCCCCC(=O)ON2C(=O)CCC2=O

Specifications

Melting Point 166°C to 170°C
Quantity 500 mg
Solubility Information Insoluble in water. Soluble in chloroform: 50mg/mL.
Formula Weight 368.34
Percent Purity 97%
Chemical Name or Material Disuccinimidyl suberate
Hazard Category H315-H319-H335
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.