Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Dithizone, MP Biomedicals

Catalog No. icn15100001
Click to view available options
Quantity:
1 g

  • Dithizone can be used as a titrant in the quantitative determination of heavy metals
  • It is an indicator for complexometry and a reagent for the extraction of metals

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 60-10-6
Molecular Formula C13H12N4S
Molecular Weight (g/mol) 256.33
MDL Number MFCD00003025
InChI Key UOFGSWVZMUXXIY-UHFFFAOYSA-N
Synonym Diphenylthiocarbazone, Phenyldiazenecarbothioic acid 2-phenylhydrazide, (Phenylazo) thioformic acid 2-phenylhydrazide
PubChem CID 657262
IUPAC Name 3-(phenylamino)-1-(phenylimino)thiourea
SMILES S=C(NNC1=CC=CC=C1)N=NC1=CC=CC=C1

Specifications

Melting Point 168°C (decomposition)(literature)
Linear Formula C13H12N4S
Solubility Information Freely soluble in carbon tetrachloride, chloroform. Sparingly soluble in alcohols. Insoluble in water. Soluble in pyridine (10mg/mL -clear brown to dark green solution).
Formula Weight 256.33
Percent Purity ≥85%
Color Black
Quantity 1 g
Physical Form Powder
Chemical Name or Material DITHIZONE
Recommended Storage Store at Room Temperature (15-30°C).
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.