Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Dithranol, 99%, MP Biomedicals™

Catalog No. p-4405601
Click to view available options
Quantity:
0.25 g
1 g
5 g

Dithranol ≥99%, Yellow Powder

  • Dithranol, a potential anti-psoriasis medication, may be used to study its safety, efficacy, metabolism and pharmacological profile.
  • It may be used to study is physicochemical properties, as a reference compound and to develop effective drug delivery vehicles.
  • It may be used as a matrix for matrix assisted laser desorption/ionization imaging on a Fourier transform ion cyclotron resonance mass spectrometer.
  • Anthralin is a leukotriene biosynthesis inhibitor.
  • Inhibits LTB4 ω-oxidation and disrupts mitochondria function.
  • Anthralin is used in the treatment of psoriasis, as a fungicide, in the treatment of ringworm infections and in chronic dermatoses.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1143-38-0
Molecular Formula C14H10O3
Molecular Weight (g/mol) 226.23
MDL Number MFCD00053409
InChI Key NUZWLKWWNNJHPT-UHFFFAOYSA-N
PubChem CID 2202
ChEBI CHEBI:37510
IUPAC Name 1,8-dihydroxy-9,10-dihydroanthracen-9-one
SMILES OC1=CC=CC2=C1C(=O)C1=C(O)C=CC=C1C2

Specifications

Color Orange-Yellow or Yellow
Assay Percent Range ≥99%
Quantity 0.25 g
Synonym anthralin, dithranol, 1,8-dihydroxyanthrone, 1,8-dihydroxy-9-anthrone, chrysodermol, cigthranol, batridol, cignolin, psoriacid-stift, 9 10h-anthracenone, 1,8-dihydroxy
Formula Weight 226.23
Physical Form Crystalline Powder
Chemical Name or Material Dithranol
EINECSNumber 214-538-0
Recommended Storage Store at Room Temperature (15°to 30°C)
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.