Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Dithranol, BP, 98.5-101%, Spectrum™ Chemical
SDP

Catalog No. 18602270 Shop All Spectrum Chemical Manufacturing Corporation Products
Click to view available options
Quantity:
10 g
25 g
100 g
1 kg
5 kg
Packaging:
Amber Glass Bottle

AN126, 1143-38-0, C14H10O3

Spectrum™ Chemical Dithranol, BP is used to treat chronic plaque psoriasis. Spectrum Chemical manufactured BP products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

CAS 1143-38-0
Molecular Formula C14H10O3
Molecular Weight (g/mol) 226.23
MDL Number MFCD00053409
InChI Key NUZWLKWWNNJHPT-UHFFFAOYSA-N
IUPAC Name 1,8-dihydroxy-9,10-dihydroanthracen-9-one
SMILES OC1=CC=CC2=C1C(=O)C1=C(O)C=CC=C1C2

Specifications

CAS Max % 100%
Loss on Drying 0.5%
Packaging Amber Glass Bottle
Quantity 10 g
Percent Purity 98.5 to 101%
Grade BP
Physical Form Crystalline Powder or Lumps
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.