Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Diversey™ Diton™ B
Click to view available options
Volume (English):
8 oz.
Description
- Cleaning powder for food and beverage industries
Specifications
Specifications
Product Type | Diton™ B |
Form | Powder |
Sterility | Non-sterile |
Appearance | White |
Filtered | Non-filtered |
For Use With (Application) | Manual cleaner for professional cleaning maintenance in food and beverage industries |
Quantity | 100 Pouches |
Volume (English) | 8 oz. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction