Learn More
Dityrosine Antibody (10A6), FITC, Novus Biologicals™
SDP
Mouse Monoclonal Antibody
Supplier:  Novus Biologicals NBP259361F
Catalog No. NBP259361F
Only null left
Add to Cart
Description
Description
Dityrosine Monoclonal specifically detects Dityrosine in Non-species specific samples. It is validated for Flow Cytometry, ELISA, Immunocytochemistry/Immunofluorescence.
Specifications
Specifications
Dityrosine
Monoclonal
FITC
PBS with 0.05% Sodium Azide
Synthetic Dityrosine conjugated to Keyhole Limpet Hemocyanin (KLH).
100 μL
Specific for dityrosine modified proteins. Does not cross-react with 3,5-dibromotyrosine or bromotyrosine modified proteins.
Store at 4C in the dark.
IgG1
Flow Cytometry, ELISA, Immunocytochemistry, Immunofluorescence
10A6
Flow Cytometry, ELISA, Immunocytochemistry/Immunofluorescence
Mouse
Protein G purified
Primary
Non-species specific
Purified
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Product Certifications
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.