Learn More
Invitrogen™ Dityrosine Monoclonal Antibody (7D4), PerCP
Mouse Monoclonal Antibody
Supplier: Invitrogen™ MA545827
Description
A 1:1,000 dilution of MA5-45827 was sufficient for detection of dityrosine in 1 μg of Dityrosine conjugated to BSA by ECL immunoblot analysis using Goat Anti-Mouse IgG:HRP as the secondary Antibody. Specific for dityrosine modified proteins. Does not cross-react with 3,5-dibromotyrosine or bromotyrosine modified proteins.
Dityrosine may be a natural component of protein structure, a product of environmental stress, or a product of in vitro protein modification.Dityrosine is involved in diverse reactions that produce tyrosyl radicals, which in turn may couple to yield dityrosine.
Specifications
Dityrosine | |
Monoclonal | |
1 mg/mL | |
2.48mM MES, 95.64mM Phosphate with 0.5M EDTA and no preservative; pH 7.4 | |
Mouse | |
Protein G | |
RUO | |
Chemical | |
Antibody | |
IgG1 |
ELISA, Western Blot, Immunocytochemistry | |
7D4 | |
PerCP | |
Dityrosine; Dityrosine (DT); DT; DY | |
Synthetic Dityrosine conjugated to Keyhole Limpet Hemocyanin (KLH). | |
100 μg | |
Primary | |
4°C | |
Liquid | |
Chemical |
Safety and Handling
Your input is important to us. Please complete this form to provide feedback related to the content on this product.