Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Divalproex Sodium, USP, 98.10-102%, Spectrum™ Chemical
SDP

Catalog No. 18602279
Click to view available options
Quantity:
25 g
100 g
500 g
1 kg
5 kg
Packaging:
Amber Glass Bottle
Poly Pail

D2044, 76584-70-8, (C16H31NaO4)n

Spectrum™ Chemical Divalproex Sodium, USP is used to treat seizure disorders, certain psychiatric conditions (manic phase of bipolar disorder), and to prevent migraine headaches. All Spectrum Chemical USP grade products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

CAS 76584-70-8
Molecular Formula C16H31NaO4
Molecular Weight (g/mol) 310.41
InChI Key MSRILKIQRXUYCT-UHFFFAOYSA-M
IUPAC Name sodium 2-propylpentanoic acid 2-propylpentanoate
SMILES [Na+].CCCC(CCC)C(O)=O.CCCC(CCC)C([O-])=O

Specifications

CAS Max % 100%
Packaging Poly Pail
Quantity 5 kg
Percent Purity 98.10 to 102%
Grade USP
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.