Learn More
GSC Go Science Crazy Diving Bell Demonstration
Shop All GSC Products
Click to view available options
:
Diving bell demonstration kit
Description
- Visually demonstrates buoyancy
- Includes Bell, syringe, connecting tube, and instructions
Specifications
Specifications
| Product Type | Diving bell demonstration kit |
| Includes | Bell, syringe, connecting tube, instructions |
| For Use With (Application) | To understand air pressure properties, density and Archimedes principle |
Safety and Handling
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov