Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Divinyl Adipate (stabilized with MEHQ) 99.0+%, TCI America™
SDP

Catalog No. A1188100G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A1188100G 100 g
A118825G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A1188100G Supplier TCI America Supplier No. A1188100G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 4074-90-2
Molecular Formula C10H14O4
Molecular Weight (g/mol) 198.22
MDL Number MFCD00059202
InChI Key JZQAAQZDDMEFGZ-UHFFFAOYSA-N
Synonym Adipic Acid Divinyl Ester
PubChem CID 19997
IUPAC Name 1,6-diethenyl hexanedioate
SMILES C=COC(=O)CCCCC(=O)OC=C

Specifications

Color White
Quantity 100 g
Formula Weight 198.22
Percent Purity ≥99.0% (GC)
Physical Form Crystalline Lumps
Chemical Name or Material Divinyl Adipate (stabilized with MEHQ)
EINECSNumber (2)-0870
RTECSNumber AV1800000
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.