Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Divinylbenzene

Catalog No. 169099250ML Shop All MilliporeSigma Sigma Organic Chemistry Products
Click to view available options
Quantity:
250 mL
1 L

Catalog No. 16-909-9250ML

Supplier: Sigma Aldrich 169099250ML

Only null left
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Divinylbenzene (DVB) has a benzene ring in combination with meta and para isomers of DVB and ethylvinylbenzene. It acts as a monomeric unit which facilitates the preparation of polydivinylbenzene by anionic polymerization.DVB can be used as a crosslinking agent in the synthesis of silicon/carbon composite for use as an anode material for lithium ion batteries. It can also be used as a monomer to synthesize dimer particles by seed polymerization method for potential uses in electrical clutches and shock absorbers.contains 4-ethylvinylbenzene and 3-ethylvinylbenzene

Specifications

CAS 1321-74-0
Density 0.912 g/mL (at 25°C)
Boiling Point 195°C
Molecular Formula C10H10
Refractive Index n20/D 1.561 (literature)
Linear Formula C6H4(CH=CH2)2
Quantity 250 mL
MDL Number MFCD01778656
Synonym DVB HP; Diethenylbenzene; Vinylstyrene
Molecular Weight (g/mol) 130.19
Percent Purity 55%
Biological Activity Respiratory system
Show More Show Less
Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.