Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Divinylbenzene, 80%, mixture of isomers, stab. with 1000ppm 4-tert-butylcatechol

Catalog No. AA4280436
Click to view available options
Quantity:
25 g
100 g
500 g

CAS: 1321-74-0 | C10H10 | 130.19 g/mol

Divinylbenzene is used as a reactive monomer in polyester resins and for the synthesis of highly crosslinked macroporous styrene as well as a crosslinking agent. It acts as an intermediate in the production of ion exchange resin. Further, it is used in making rubber, petroleum production and polymeric core shell nanoparticles. In addition, it is used to synthesize monodispersed poly divinylbenzene microspheres.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1321-74-0
Molecular Formula C10H10
Molecular Weight (g/mol) 130.19
MDL Number MFCD01778656,MFCD00010654
InChI Key MYRTYDVEIRVNKP-UHFFFAOYSA-N
Synonym divinylbenzene, 1,2-divinylbenzene, o-divinylbenzene, benzene, diethenyl, benzene, 1,2-diethenyl, divinylbenzen, divinyl benzen, divinyl-benzene, poly divinylbenzene, divinylbenzene dvb
PubChem CID 66666
IUPAC Name 1,2-bis(ethenyl)benzene
SMILES C=CC1=CC=CC=C1C=C

Specifications

Color Colorless to Yellow
Density 0.914 g/mL
Flash Point 69°C (156°F)
Refractive Index 1.574
Quantity 500 g
Sensitivity Light sensitive
Solubility Information Miscible with ethanol and ether. Immiscible with water.
Formula Weight 130.19
Percent Purity 80%
Physical Form Liquid
Chemical Name or Material Divinylbenzene, Mixture of isomers, Stabilized with 1000ppm 4-tert-butylcatechol
Show More Show Less
Hazard Category H227-H315-H319
Hazard Statement GHS H Statement
H315-H319-H335-H227
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Combustible liquid.
Precautionary Statement P210-P264b-P280-P302+P352-P305+P351+P338-P332+P313-P362-P370+P378q-P501c
EINECSNumber 215-325-5
RTECSNumber CZ9370000
TSCA Yes
Recommended Storage Keep cold

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.