Learn More
DIXDC1 Mouse anti-Human, Clone: OTI1A2, lyophilized, TrueMAB™

Mouse Monoclonal Antibody
Supplier: OriGene CF505521

Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
The protein encoded by this gene is a positive regulator of the Wnt signaling pathway. The encoded protein is found associated with gamma tubulin at the centrosome. Alternative splicing results in multiple transcript variants encoding different isoforms.Specifications
DIXDC1 | |
Monoclonal | |
Unconjugated | |
DIXDC1 | |
CCD1 | |
Mouse | |
Affinity Chromatography | |
RUO | |
85458 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Lyophilized |
Western Blot | |
OTI1A2 | |
PBS with 8% trehalose and no preservative; pH 7.3 | |
Q155Q3 | |
DIXDC1 | |
Full length human recombit protein of human DIXDC1 produced in HEK293T cell. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG1 |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.