Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ DIXDC1 Polyclonal Antibody
Goat Polyclonal Antibody
Supplier: Invitrogen™ PA519232
Description
This antibody is predicted to react with bovine, canine, mouse and rat based on sequence homology. This antibody is tested in Peptide ELISA: antibody detection limit dilution 64,000.
This gene encodes one of four subunits of the splicing factor 3B. The protein encoded by this gene cross-links to a region in the pre-mRNA immediately upstream of the branchpoint sequence in pre-mRNA in the prespliceosomal complex A. It also may be involved in the assembly of the B, C and E spliceosomal complexes. In addition to RNA-binding activity, this protein interacts directly and highly specifically with subunit 2 of the splicing factor 3B. This protein contains two N-terminal RNA-recognition motifs (RRMs), consistent with the observation that it binds directly to pre-mRNA.
Specifications
DIXDC1 | |
Polyclonal | |
Unconjugated | |
Dixdc1 | |
4930563F16Rik; BC048182; ccd1; Ccd1Aalpha1L; Ccd1Abeta1L; Ccd1BalphaL; Ccd1BbetaL; Ccd1CL; coiled-coil DIX 1; coiled-coil protein DIX1; Coiled-coil protein DIX1-A; coiled-coil-DIX1; Coiled-coil-DIX1-A; DIX domain containing 1; DIX domain containing 1a; DIX domain-containing protein 1; DIX domain-containing protein 1-A; Dixdc1; dixdc1a; Dixin; dixin-A; EGK_06890; Kiaa1735; mKIAA1735 | |
Goat | |
Ammonium sulfate precipitation | |
RUO | |
85458 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
0.5 mg/mL | |
TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3 | |
Q155Q3 | |
Dixdc1 | |
Synthetic peptide sequence (DVSYHQVDLERE) corresponding to the internal amino acids of DIXDC1 (aa 450-462). | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction