Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ DIXDC1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA565243
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA565243 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA565243 Supplier Invitrogen™ Supplier No. PA565243
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: KSESIITQSEE KADFVIIPAE GIENRTEGTD SPLSRDWRPG SPGTYLETSW EEQLLEQQEY LEKEMEEAKK MISGLQALLL NGSLPEDEQE RPLALCEPGV NP Highest antigen sequence identity to the following orthologs - mouse 89%, rat 92%.

This gene encodes one of four subunits of the splicing factor 3B. The protein encoded by this gene cross-links to a region in the pre-mRNA immediately upstream of the branchpoint sequence in pre-mRNA in the prespliceosomal complex A. It also may be involved in the assembly of the B, C and E spliceosomal complexes. In addition to RNA-binding activity, this protein interacts directly and highly specifically with subunit 2 of the splicing factor 3B. This protein contains two N-terminal RNA-recognition motifs (RRMs), consistent with the observation that it binds directly to pre-mRNA.
TRUSTED_SUSTAINABILITY

Specifications

Antigen DIXDC1
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.4 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene Dixdc1
Gene Accession No. Q155Q3
Gene Alias 4930563F16Rik; BC048182; ccd1; Ccd1Aalpha1L; Ccd1Abeta1L; Ccd1BalphaL; Ccd1BbetaL; Ccd1CL; coiled-coil DIX 1; coiled-coil protein DIX1; Coiled-coil protein DIX1-A; coiled-coil-DIX1; Coiled-coil-DIX1-A; DIX domain containing 1; DIX domain containing 1a; DIX domain-containing protein 1; DIX domain-containing protein 1-A; Dixdc1; dixdc1a; Dixin; dixin-A; EGK_06890; Kiaa1735; mKIAA1735
Gene Symbols Dixdc1
Host Species Rabbit
Immunogen Recombinant Human DIXDC1. Recombinant protein control fragment (Product #RP-104706).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 85458
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.