Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ DIXDC1A Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA572666
Description
DIXDC1A Polyclonal Antibody for Western Blot
Functions as a positive effector of the Wnt signaling pathway regulating antero-posterior neural patterning.
Specifications
DIXDC1A | |
Polyclonal | |
Unconjugated | |
dixdc1a | |
ccd1; coiled-coil DIX 1; coiled-coil protein DIX1; coiled-coil protein DIX1-A; coiled-coil-DIX1; coiled-coil-DIX1-A; DIX domain containing 1; DIX domain containing 1a; DIX domain-containing protein 1; DIX domain-containing protein 1-A; dixdc1; dixdc1a; dixin; dixin-A | |
Rabbit | |
Antigen affinity chromatography, Protein A | |
RUO | |
360137 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Western Blot | |
0.6 mg/mL | |
PBS with 0.09% sodium azide; pH 7.4 | |
Q804T6 | |
dixdc1a | |
KLH conjugated synthetic peptide between 282-316 amino acids of DANRE dixdc1a. | |
400 μL | |
Primary | |
Zebrafish | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction