Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ DJ-1 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5143602
Description
DJ-1 Polyclonal Antibody for Western Blot, ICC/IF, IHC
The product of this gene belongs to the peptidase C56 family of proteins. It acts as a positive regulator of androgen receptor-dependent transcription. It may also function as a redox-sensitive chaperone, as a sensor for oxidative stress, and it apparently protects neurons against oxidative stress and cell death. Defects in this gene are the cause of autosomal recessive early-onset Parkinson disease 7. Two transcript variants encoding the same protein have been identified for this gene.
Specifications
DJ-1 | |
Polyclonal | |
Unconjugated | |
Park7 | |
CAP1; Contraception-associated protein 1; CTA-215D11.1; dj1; DJ-1; DJ-1 protein; early onset 7; epididymis secretory sperm binding protein Li 67p; fertility protein SP22; FLJ27376; FLJ34360; FLJ92274; HEL-S-67p; hypothetical protein LOC449674; maillard deglycase; Oncogene DJ1; OTTHUMP00000001350; OTTHUMP00000001351; PARK7; PARK7 GENE; PARK7/DJ1; parkinson disease (autosomal recessive, early onset) 7; Parkinson disease 7; Parkinson disease protein 7; Parkinson disease protein 7 homolog; Parkinson disease protein 7 homolog; protein/nucleic acid deglycase DJ-1; parkinson protein 7; Parkinsonism associated deglycase; parkinsonism-associated deglycase; Protein CAP1; protein deglycase DJ-1; protein deglycase DJ-1zDJ-1; Protein DJ-1; protein DJ-1-like protein; protein DJ-1zDJ-1; Protein/nucleic acid deglycase DJ-1; RNA-binding protein regulatory subunit; SP22; zDJ-1; zgc:103725 | |
Rabbit | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Immunohistochemistry, Western Blot, Immunocytochemistry | |
Conc. Not Determined | |
whole serum with 5mM sodium azide | |
O88767, Q99497, Q99LX0 | |
Park7 | |
Full length recombinant human DJ1 expressed in and purified from E. coli. | |
RUO | |
11315, 117287, 57320 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction