Learn More
Invitrogen™ DJ-1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5120650
Description
Positive test controls include: HeLa, SH-SY5Y, BxPC-3, HepG2, Mouse thymus, Mouse kidney, Mouse pancreas, Rat brain, Rat testis. The target is usually found in the following locations: Cell membrane, Cytoplasm, Lipid-anchor, Membrane raft, Mitochondrion, Nucleus. Immunogen sequence: MASKRALVIL AKGAEEMETV IPVDVMRRAG IKVTVAGLAG KDPVQCSRDV VICPDASLED AKKEGPYDVV VLPGGNLGAQ NLSESAAVKE ILKEQENRKG LIAAICAGPT ALLAHEIGFG SKVTTHPLAK DKMMNGGHYT YSENRVEKDG LILTSRGPGT SFEFALAIVE ALNGKEVAAQ VKAPLVLKD.
The product of this gene belongs to the peptidase C56 family of proteins. It acts as a positive regulator of androgen receptor-dependent transcription. It may also function as a redox-sensitive chaperone, as a sensor for oxidative stress, and it apparently protects neurons against oxidative stress and cell death. Defects in this gene are the cause of autosomal recessive early-onset Parkinson disease 7. Two transcript variants encoding the same protein have been identified for this gene.
Specifications
| DJ-1 | |
| Polyclonal | |
| Unconjugated | |
| Park7 | |
| CAP1; Contraception-associated protein 1; CTA-215D11.1; dj1; DJ-1; DJ-1 protein; early onset 7; epididymis secretory sperm binding protein Li 67p; fertility protein SP22; FLJ27376; FLJ34360; FLJ92274; HEL-S-67p; hypothetical protein LOC449674; maillard deglycase; Oncogene DJ1; OTTHUMP00000001350; OTTHUMP00000001351; PARK7; PARK7 GENE; PARK7/DJ1; parkinson disease (autosomal recessive, early onset) 7; Parkinson disease 7; Parkinson disease protein 7; Parkinson disease protein 7 homolog; Parkinson disease protein 7 homolog; protein/nucleic acid deglycase DJ-1; parkinson protein 7; Parkinsonism associated deglycase; parkinsonism-associated deglycase; Protein CAP1; protein deglycase DJ-1; protein deglycase DJ-1zDJ-1; Protein DJ-1; protein DJ-1-like protein; protein DJ-1zDJ-1; Protein/nucleic acid deglycase DJ-1; RNA-binding protein regulatory subunit; SP22; zDJ-1; zgc:103725 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 11315, 117287, 57320 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ELISA, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 1.63 mg/mL | |
| PBS with 50% glycerol and 0.05% ProClin 300; pH 7.3 | |
| O88767, Q99497, Q99LX0 | |
| Park7 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-189 of human DJ-1/PARK7 (NP_009193.2). | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.