Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DJ-1 Polyclonal Antibody, Invitrogen™

Catalog No. PIPA132440
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA132440 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA132440 Supplier Thermo Scientific Supplier No. PA132440

Rabbit Polyclonal Antibody

PA1-32440 detects DJ1 from mouse, human, rat, hamster and fish samples.

The product of this gene belongs to the peptidase C56 family of proteins. It acts as a positive regulator of androgen receptor-dependent transcription. It may also function as a redox-sensitive chaperone, as a sensor for oxidative stress, and it apparently protects neurons against oxidative stress and cell death. Defects in this gene are the cause of autosomal recessive early-onset Parkinson disease 7. Two transcript variants encoding the same protein have been identified for this gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen DJ-1
Applications Immunocytochemistry, Immunohistochemistry (Paraffin), Immunoprecipitation, In vitro Assay, Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation whole serum with no preservative
Gene Park7
Gene Accession No. O88767, Q5E946, Q5XJ36, Q8UW59, Q99497, Q99LX0
Gene Alias CAP1; Contraception-associated protein 1; CTA-215D11.1; dj1; DJ-1; DJ-1 protein; early onset 7; epididymis secretory sperm binding protein Li 67p; fertility protein SP22; FLJ27376; FLJ34360; FLJ92274; HEL-S-67p; hypothetical protein LOC449674; maillard deglycase; Oncogene DJ1; OTTHUMP00000001350; OTTHUMP00000001351; PARK7; PARK7 GENE; PARK7/DJ1; parkinson disease (autosomal recessive, early onset) 7; Parkinson disease 7; Parkinson disease protein 7; Parkinson disease protein 7 homolog; Parkinson disease protein 7 homolog; protein/nucleic acid deglycase DJ-1; parkinson protein 7; Parkinsonism associated deglycase; parkinsonism-associated deglycase; Protein CAP1; protein deglycase DJ-1; protein deglycase DJ-1zDJ-1; Protein DJ-1; protein DJ-1-like protein; protein DJ-1zDJ-1; Protein/nucleic acid deglycase DJ-1; RNA-binding protein regulatory subunit; SP22; zDJ-1; zgc:103725
Gene Symbols Park7
Host Species Rabbit
Immunogen Synthetic peptide corresponding to residues between 150-189 of human DJ-1 (the C-terminus).
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 100759195, 11315, 117287, 395277, 449674, 511268, 57320
Target Species Bovine, Chicken, Fish, Hamster, Human, Mouse, Rat, Zebrafish
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.