Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ DJC18 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA557707
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA557707 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA557707 Supplier Invitrogen™ Supplier No. PA557707
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: STLGYTISRE TQNLQVPYFV DKNFDKAYRG ASLHDLEKTI EKDYIDYIQT SCWKEKQQKS ELTNLAGLYR DERLKQKAES LKLENCEKLS KL Highest antigen sequence identity to the following orthologs: Mouse - 97%, Rat - 97%.

DNAJC18 gene ontology annotations related to this gene include cellular response to misfolded protein; chaperone cofactor-dependent protein refolding; ubiquitin-dependent ERAD pathway.
TRUSTED_SUSTAINABILITY

Specifications

Antigen DJC18
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene DNAJC18
Gene Accession No. Q9H819
Gene Alias DnaJ (Hsp40) homolog, subfamily C, member 18; DnaJ heat shock protein family (Hsp40) member C18; dnaJ homolog subfamily C member 18; DNAJC18; RGD1310237
Gene Symbols DNAJC18
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human DJC18. Recombinant protein control fragment (Product #RP-96932).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 202052
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.