Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ DJC18 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA566307
Description
Immunogen sequence: TPEELFNVFFG GHFPTGNIHM FSNVTDDTYY YRRRHRHERT QTQKEEEEEK PQTTYSA Highest antigen sequence identity to the following orthologs - mouse 83%, rat 81%.
DNAJC18 gene ontology annotations related to this gene include cellular response to misfolded protein; chaperone cofactor-dependent protein refolding; ubiquitin-dependent ERAD pathway.
Specifications
DJC18 | |
Polyclonal | |
Unconjugated | |
DNAJC18 | |
DnaJ (Hsp40) homolog, subfamily C, member 18; DnaJ heat shock protein family (Hsp40) member C18; dnaJ homolog subfamily C member 18; DNAJC18; RGD1310237 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
202052 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry | |
0.1 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q9H819 | |
DNAJC18 | |
Recombinant Human DJC18. Recombinant protein control fragment (Product #RP-106985). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction