Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ DJC18 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5116777
Description
Positive Samples: BxPC-3, Jurkat Immunogen sequence: STLGYTISRE TQNLQVPYFV DKNFDKAYRG ASLHDLEKTI EKDYIDYIQT SCWKEKQQKS ELTNLAGLYR DERLKQKAES LKLENCEKLS K.
DNAJC18 gene ontology annotations related to this gene include cellular response to misfolded protein; chaperone cofactor-dependent protein refolding; ubiquitin-dependent ERAD pathway.
Specifications
DJC18 | |
Polyclonal | |
Unconjugated | |
DNAJC18 | |
DnaJ (Hsp40) homolog, subfamily C, member 18; DnaJ heat shock protein family (Hsp40) member C18; dnaJ homolog subfamily C member 18; DNAJC18; RGD1310237 | |
Rabbit | |
Affinity chromatography | |
RUO | |
202052, 291677 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
ELISA, Western Blot | |
3.04 mg/mL | |
PBS with 50% glycerol and 0.01% thimerosal; pH 7.3 | |
Q9H819 | |
DNAJC18 | |
Recombinant fusion protein containing a sequence corresponding to amino acids 260-350 of human DNAJC18 (NP_6898991). | |
100 μL | |
Primary | |
Human, Rat | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction