Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
DjC9 Antibody, DyLight 650, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NB10097848C
Description
DjC9 Polyclonal antibody specifically detects DjC9 in Human samples. It is validated for Western BlotSpecifications
DjC9 | |
Polyclonal | |
Western Blot | |
DnaJ (Hsp40) homolog, subfamily C, member 9, dnaJ homolog subfamily C member 9 | |
The immunogen recognized by this antibody maps to a region between residue 210 and 260 of human DnaJ (Hsp40) homolog, subfamily C, member 9 using the numbering given in entry NP_056005.12 (GeneID 23234). | |
0.1 mL | |
Signal Transduction | |
23234 | |
Store at 4C in the dark. | |
IgG |
Western Blot | |
DyLight 650 | |
50mM Sodium Borate | |
Rabbit | |
Immunogen affinity purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction