Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
DjC9 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NB10097848
Description
DjC9 Polyclonal specifically detects DjC9 in Human samples. It is validated for Western Blot, Immunoprecipitation.Specifications
DjC9 | |
Polyclonal | |
Unconjugated | |
Tris-Citrate/Phosphate (pH 7.0 - 8.0) with 0.09% Sodium Azide | |
DNAJC9 | |
The immunogen recognized by this antibody maps to a region between residue 210 and 260 of human DnaJ (Hsp40) homolog, subfamily C, member 9 using the numbering given in entry NP_056005.12 (GeneID 23234). | |
100 μL | |
Primary | |
Human | |
IgG |
Western Blot, Immunoprecipitation | |
1.0 mg/mL | |
Western Blot 1:2000-1:10000, Immunoprecipitation 1:10-1:500 | |
DnaJ (Hsp40) homolog, subfamily C, member 9, dnaJ homolog subfamily C member 9 | |
Rabbit | |
Affinity Purified | |
RUO | |
23234 | |
Store at 4C. Do not freeze. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction