Learn More
DjC9 Rabbit anti-Human, Polyclonal, Bethyl Laboratories
Rabbit Polyclonal Antibody
Supplier: Bethyl Laboratories, Inc A301840A
Description
The recommended shelf life for this product is 1 year from date of receipt.
May play a role as co-chaperone of the Hsp70 family proteins HSPA1A, HSPA1B and HSPA8.Specifications
| DjC9 | |
| Polyclonal | |
| Unconjugated | |
| DNAJC9 | |
| DnaJ homolog subfamily C member 9, DnaJ protein SB73, DNAJC9 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 23234 | |
| 4° C | |
| Liquid |
| Immunoprecipitation, Western Blot | |
| 1 mg/ml | |
| phosphate, tris citrate with 0.09% sodium azide; pH 7-8 | |
| Q8WXX5 | |
| DNAJC9 | |
| Between 210 and 260 | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.