Learn More
Invitrogen™ DKC1 Polyclonal Antibody

Description
Immunogen sequence: KERKSLPEED VAEIQHAEEF LIKPESKVAK LDTSQWPLLL KNFDKLNVRT THYTPLACGS NPLKREIGDY IRTGFINLDK PSNPSSHEVV AWIRRILRVE KTGHSGTL Highest antigen sequence indentity to the following orthologs: Mouse - 94%, Rat - 86%.
Specifications
Specifications
| Antigen | DKC1 |
| Applications | Western Blot, Immunocytochemistry |
| Classification | Polyclonal |
| Concentration | 0.10 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 |
| Gene | DKC1 |
| Gene Accession No. | O60832 |
| Gene Alias | BC068171; CBF5; CBF5 homolog; DKC; DKC1; DKCX; dyskeratosis congenita 1, dyskerin; dyskeratosis congenita 1, dyskerin homolog; dyskerin; dyskerin pseudouridine synthase 1; H/ACA ribonucleoprotein complex subunit 4; H/ACA ribonucleoprotein complex subunit DKC1; Nap57; NOLA4; Nopp140-associated protein of 57 kDa; nucleolar protein family A member 4; Nucleolar protein NAP57; snoRNP protein DKC1; XAP101 |
| Gene Symbols | DKC1 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.