Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
DKFZP434B168 Antibody, HRP, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NB100465H
Description
DKFZP434B168 Polyclonal antibody specifically detects DKFZP434B168 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)Specifications
DKFZP434B168 | |
Polyclonal | |
Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin | |
C1orf73, chromosome 1 open reading frame 73, DKFZp434B168, Int7, integrator complex subunit 7 | |
The immunogen this antibody was made to, maps to a region between residues 850-900 of human DKFZP434B168 using the TrEMBL entry Q9NVH2 (GeneID 25896). | |
0.1 mL | |
Zinc Finger | |
25896 | |
Store at 4C in the dark. | |
IgG |
Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
HRP | |
PBS | |
Rabbit | |
Immunogen affinity purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction