Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
DKFZp566F084 Antibody (OTI4B9), Alexa Fluor™ 532, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP271779AF532
Description
DKFZp566F084 Monoclonal specifically detects DKFZp566F084 in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence.Specifications
DKFZp566F084 | |
Monoclonal | |
Alexa Fluor 532 | |
50 mM sodium borate with 0.05% sodium azide | |
DPCD | |
Full length recombinant protein of human DKFZp566F084 (NP_056263) produced in HEK293T cells. | |
Primary | |
Human, Mouse | |
IgG1 |
Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence | |
OTI4B9 | |
Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence | |
deleted in a mouse model of primary ciliary dyskinesia, deleted in primary ciliary dyskinesia homolog (mouse), DKFZP566F084, protein DPCD, RP11-529I10.4 | |
Mouse | |
0.1 mL | |
25911 | |
Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction