Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Dkk-3 Antibody (019), Alexa Fluor™ 488, Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP290496AF488
Description
Dkk-3 Monoclonal specifically detects Dkk-3 in Mouse samples. It is validated for ELISA.Specifications
Dkk-3 | |
Monoclonal | |
Alexa Fluor 488 | |
dickkopf (Xenopus laevis) homolog 3, dickkopf 3, dickkopf homolog 3 (Xenopus laevis), Dickkopf-3, dickkopf-related protein 3, Dkk-3, hDkk-3, regulated in glioma, REICdkk-3, RIG, RIG-like 5-6, RIG-like 7-1 | |
This antibody was obtained from a rabbit immunized with purified, recombinant Mouse Dkk-3 (NP_056629.1; Met 1-Ile 349). | |
0.1 mL | |
Signal Transduction | |
27122 | |
Mouse | |
Purified |
ELISA | |
19 | |
50 mM Sodium Borate | |
Rabbit | |
Protein A purified | |
RUO | |
Primary | |
No cross-reactivity in ELISA with: Human DKK3 | |
Store at 4°C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction