Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Dkk-3 Rabbit anti-Human, Mouse, Rat, Clone: 1R5E0, Novus Biologicals™

Rabbit Monoclonal Antibody
$205.50 - $458.50
Specifications
Antigen | Dkk-3 |
---|---|
Clone | 1R5E0 |
Applications | Western Blot, Immunofluorescence |
Classification | Monoclonal |
Conjugate | Unconjugated |
Description
Dkk-3 Monoclonal antibody specifically detects Dkk-3 in Human, Mouse, Rat samples. It is validated for Western Blot, ImmunofluorescenceSpecifications
Dkk-3 | |
Western Blot, Immunofluorescence | |
Unconjugated | |
Rabbit | |
Signal Transduction | |
PBS, 0.05% BSA, 50% glycerol, pH7.3 | |
27122 | |
IgG | |
Affinity purified |
1R5E0 | |
Monoclonal | |
Purified | |
RUO | |
Human, Mouse, Rat | |
dickkopf (Xenopus laevis) homolog 3, dickkopf 3, dickkopf homolog 3 (Xenopus laevis), Dickkopf-3, dickkopf-related protein 3, Dkk-3, hDkk-3, regulated in glioma, REICdkk-3, RIG, RIG-like 5-6, RIG-like 7-1 | |
Recombinant fusion protein containing a sequence corresponding to amino acids 200-360 of human Dkk-3 (Q9UBP4). | |
Primary | |
Store at -20°C. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title