Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human Dkk-4 Antibody, R&D Systems™

Goat Polyclonal Antibody
$200.50 - $549.50
Specifications
Antigen | Dkk-4 |
---|---|
Concentration | LYOPH |
Dilution | Western Blot 0.1 ug/mL |
Applications | Western Blot |
Classification | Polyclonal |
Description
Dkk-4 Polyclonal specifically detects Dkk-4 in Human samples. It is validated for Western Blot.Specifications
Dkk-4 | |
Western Blot 0.1 ug/mL | |
Polyclonal | |
Goat | |
Human | |
dickkopf (Xenopus laevis) homolog 4, dickkopf homolog 4 (Xenopus laevis), dickkopf-4, dickkopf-related protein 4, Dkk4, DKK-4, hDkk-4, MGC129562, MGC129563 | |
DKK4 | |
IgG | |
Affinity Purified | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
LYOPH | |
Western Blot | |
Unconjugated | |
RUO | |
Q9UBT3 | |
27121 | |
Mouse myeloma cell line NS0-derived recombinant human Dkk-4 Leu19-Leu224 Accession # Q9UBT3 | |
Primary | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Detects human Dkk-4 in direct ELISAs and Western blots. In direct ELISAs, less than 1% cross-reactivity with recombinant human (rh) Dkk-1, rhDkk-3, and recombinant mouse Dkk-3 is observed. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title