Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ DKK1 Monoclonal Antibody (C5)
GREENER_CHOICE

Catalog No. PIMA541581
Change view
Click to view available options
Quantity:
20 μL
200 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA541581 200 μL
PIMA543857 20 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. PIMA541581 Supplier Invitrogen™ Supplier No. MA541581
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

DKK1 Monoclonal Antibody for Western Blot

DKK-1 is a member of the DKK protein family which also includes DKK-2, DKK-3 and DKK-4. DKK-1 was originally identified as a Xenopus head-forming molecule that behaves as an antagonist for Wnt signaling. Subsequent studies have shown that DKK-1 and DKK-4 play important regulatory roles in the Wnt/beta-catenin signaling pathway by forming inhibitory complexes with LDL receptor-related proteins 5 and 6 (LRP5 and LRP6), which are essential components of the Wnt/beta-catenin signaling system. LRP5 and LRP6 are single-pass transmembrane proteins that appear to act as co-receptors for Wnt ligands involved in the Wnt/beta-catenin signaling cascade. It has been suggested that by inhibiting Wnt/beta-catenin signaling, which is essential for posterior patterning in vertebrates, DKK-1 permits anterior development. This notion is supported by the finding that mice deficient of DKK-1 expression lack head formation and die during embryogenesis.
TRUSTED_SUSTAINABILITY

Specifications

Antigen DKK1
Applications Western Blot
Classification Monoclonal
Clone C5
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.05% ProClin 300; pH 7.4
Gene DKK1
Gene Accession No. O54908, O94907
Gene Alias dickkopf 1 homolog; dickkopf homolog 1; dickkopf homolog 1 (Xenopus laevis); dickkopf related protein-1; dickkopf WNT signaling pathway inhibitor 1; Dickkopf1; dickkopf-1; dickkopf-1 like; dickkopf-like protein 1; dickkopf-related protein 1; DKK1; DKK-1; hDkk1; hDkk-1; mdkk-1; SK; UNQ492/PRO1008
Gene Symbols DKK1
Host Species Mouse
Immunogen Recombinant protein Dickkopf Related Protein 1. The antigen corresponds to amino acid range 33-266 of the target protein.
Purification Method Protein A/G
Quantity 200 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 13380, 22943
Target Species Human, Mouse
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG2b κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.