Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
DKK2 Monoclonal Antibody (C1), Invitrogen™
Mouse Monoclonal Antibody
$221.50 - $541.50
Specifications
Antigen | DKK2 |
---|---|
Clone | C1 |
Concentration | 1 mg/mL |
Applications | Western Blot |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
PIMA543856
|
Invitrogen™
MA543856 |
20 μL |
Each for $221.50
|
|
|||||
PIMA543855
|
Invitrogen™
MA543855 |
100 μL |
Each for $541.50
|
|
|||||
Description
This gene encodes a protein that is a member of the dickkopf family. The secreted protein contains two cysteine rich regions and is involved in embryonic development through its interactions with the Wnt signaling pathway. It can act as either an agonist or antagonist of Wnt/beta-catenin signaling, depending on the cellular context and the presence of the co-factor kremen 2. Activity of this protein is also modulated by binding to the Wnt co-receptor LDL-receptor related protein 6.Specifications
DKK2 | |
1 mg/mL | |
Monoclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.05% ProClin 300 | |
dickkopf 2; dickkopf 2 homolog; Dickkopf gene 2; dickkopf homolog 1; dickkopf homolog 2; dickkopf homolog 2 (Xenopus laevis); dickkopf homolog 4; dickkopf related protein-2; dickkopf WNT signaling pathway inhibitor 2; dickkopf-2; Dickkopf-related protein 2; DKK2; DKK-2; Dkk4; hDkk-2; mDkk-2; UNQ682/PRO1316 | |
DKK2 | |
IgG1 κ | |
Protein A/G | |
Antibody |
C1 | |
Western Blot | |
Unconjugated | |
Mouse | |
Human | |
Q9UBU2 | |
27123 | |
Ile38-Lys258 (Accession Q9UBU2), with N-terminal His Tag. | |
Primary | |
-20°C | |
DKK2 |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title