Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ DKK2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA511616
Change view
Click to view available options
Quantity:
400 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA511616 400 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA511616 Supplier Invitrogen™ Supplier No. PA511616
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

DKK2 Polyclonal Antibody for Western Blot, IHC (P)

This gene encodes a protein that is a member of the dickkopf family. The secreted protein contains two cysteine rich regions and is involved in embryonic development through its interactions with the Wnt signaling pathway. It can act as either an agonist or antagonist of Wnt/beta-catenin signaling, depending on the cellular context and the presence of the co-factor kremen 2. Activity of this protein is also modulated by binding to the Wnt co-receptor LDL-receptor related protein 6.
TRUSTED_SUSTAINABILITY

Specifications

Antigen DKK2
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1.8 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.09% sodium azide; pH 7.4
Gene DKK2
Gene Accession No. Q9QYZ8, Q9UBU2
Gene Alias dickkopf 2; dickkopf 2 homolog; Dickkopf gene 2; dickkopf homolog 1; dickkopf homolog 2; dickkopf homolog 2 (Xenopus laevis); dickkopf homolog 4; dickkopf related protein-2; dickkopf WNT signaling pathway inhibitor 2; dickkopf-2; Dickkopf-related protein 2; DKK2; DKK-2; Dkk4; hDkk-2; mDkk-2; UNQ682/PRO1316
Gene Symbols DKK2
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide between 31-62 amino acids from the N-terminal region of human DKK2.
Purification Method Size-exclusion - Dialysis, Ammonium sulfate precipitation
Quantity 400 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 27123, 56811
Target Species Human, Mouse
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.