Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ DKK3 Recombinant Rabbit Monoclonal Antibody (7O2I10)
GREENER_CHOICE

Catalog No. FEMA551252
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FEMA551252 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. FEMA551252 Supplier Invitrogen™ Supplier No. MA551252
Add to Cart
Add to Cart

Rabbit Recombinant Monoclonal Antibody

Immunogen sequence: APAPTATSAP VKPGPALSYP QEEATLNEMF REVEELMEDT QHKLRSAVEE MEAEEAAAKA SSEVNLANLP PSYHNETNTD TKVGNNTIHV HREIHKITNN QTGQMVFSET VITSVGDEEG RRSHECIIDE DCGPSMYCQF ASFQYTCQPC RGQRMLCTRD SECCGDQLCV WGHCTKMAT.

DKK3, like DKK1, DKK2, and DKK4, possesses an N-terminal signal peptide and 2 conserved cysteine-rich domains, which are separated by a linker region and contain 10 cysteine residues each. The second cysteine region has a putative lipid-binding function that may facilitate WNT/DKK interactions at the plasma membrane. The linker region contains 50 to 55 amino acids in DKK1, DKK2, and DKK4, whereas in DKK3 it contains only 12 amino acids. All DKKs have several potential sites for cleavage by furin-type proteases. Northern blot analysis revealed wide expression of the DKK3 transcript, with highest expression in heart, brain, and spinal cord. In situ hybridization reveals highest expression in mouse brain, eye, and heart.
TRUSTED_SUSTAINABILITY

Specifications

Antigen DKK3
Applications ELISA, Western Blot, Immunocytochemistry
Classification Recombinant Monoclonal
Clone 7O2I10
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.05% BSA, 50% glycerol and 0.05% ProClin 300; pH 7.3
Gene DKK3
Gene Accession No. Q9QUN9, Q9UBP4
Gene Alias AW061014; C87148; Dickkopf; dickkopf 3; dickkopf 3 homolog; Dickkopf gene 3; dickkopf homolog 3; dickkopf homolog 3 (Xenopus laevis); dickkopf WNT signaling pathway inhibitor 3; Dickkopf3; dickkopf-3; Dickkopf-related protein 3; DKK 3; Dkk3; dkk-3; hDkk 3; hDkk-3; hhDkk3; mDkk-3; p29; regulated in glioma; Reic; RIG; RIG-like 5-6; RIG-like 7-1; UNQ258/PRO295; Xenopus
Gene Symbols DKK3
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 22-200 of human DKK3 (NP_056965.3).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 171548, 27122, 50781
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Format Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.