Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ DKK4 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA596812
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA596812 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA596812 Supplier Invitrogen™ Supplier No. PA596812
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: LVLDFNNIRS SADLHGARKG SQCLSDTDCN TRKFCLQPRD EKPFCATCRG LRRRCQRDAM CCPGTLCVND VCTTMEDATP ILERQLDEQD GTHAEGTTGH PVQENQPKRK PSIKKSQGRK GQEGESCLRT FDCGPGLCCA RHFWTKICKP VLLEGQVCSR RGHKDTAQAP EIFQRCDCGP GLLCRSQLTS NRQHARLRVC QKIEKL; Positive Samples: SW480, Mouse liver.

This gene encodes a protein that is a member of the dickkopf family. The secreted protein contains two cysteine rich regions and is involved in embryonic development through its interactions with the Wnt signaling pathway. Activity of this protein is modulated by binding to the Wnt co-receptor and the co-factor kremen 2.
TRUSTED_SUSTAINABILITY

Specifications

Antigen DKK4
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 0.97 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene DKK4
Gene Accession No. Q8VEJ3, Q9UBT3
Gene Alias Dickkopf gene 4; dickkopf homolog 4 (Xenopus laevis); dickkopf WNT signaling pathway inhibitor 4; dickkopf-4; Dickkopf-related protein 4; Dickkopf-related protein 4 short form; DKK4; Dkk-4; hDkk-4; MGC129562; MGC129563
Gene Symbols DKK4
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 19-224 of human DKK4 (NP_0552351).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 234130, 27121
Target Species Human, Mouse
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.