Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ DKK4 Polyclonal Antibody
Goat Polyclonal Antibody
Supplier: Invitrogen™ PA518017
Description
This antibody is predicted to react with canine, mouse, porcine and rat based on sequence homology. This antibody is tested in Peptide ELISA: antibody detection limit dilution 32,000.
This gene encodes a protein that is a member of the dickkopf family. The secreted protein contains two cysteine rich regions and is involved in embryonic development through its interactions with the Wnt signaling pathway. Activity of this protein is modulated by binding to the Wnt co-receptor and the co-factor kremen 2.
Specifications
DKK4 | |
Polyclonal | |
Unconjugated | |
DKK4 | |
Dickkopf gene 4; dickkopf homolog 4 (Xenopus laevis); dickkopf WNT signaling pathway inhibitor 4; dickkopf-4; Dickkopf-related protein 4; Dickkopf-related protein 4 short form; DKK4; Dkk-4; hDkk-4; MGC129562; MGC129563 | |
Goat | |
Ammonium sulfate precipitation | |
RUO | |
27121 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
0.5 mg/mL | |
TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3 | |
Q9UBT3 | |
DKK4 | |
Synthetic peptide sequence (RQHARLRVCQKIEKL) corresponding to the C-terminus amino acids of DKK4 (aa 210-224). | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction