Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ DKKL1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA544099
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA544099 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA544099 Supplier Invitrogen™ Supplier No. PA544099
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Peptide sequence: DALEGGHWLS EKRHRLQAIR DGLRKGTHKD VLEEGTESSS HSRLSPRKTH Sequence homology: Dog: 77%; Human: 100%; Rat: 86%.

The dickkopf protein family interacts with the Wnt signaling pathway and its members are characterized by two conserved cysteine-rich domains. This gene encodes a secreted protein that has high similarity to the N-terminus of the dickkopf-3 protein and moderate similarity to that protein's C-terminus though the C-terminal cysteine residues are not conserved.
TRUSTED_SUSTAINABILITY

Specifications

Antigen DKKL1
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene DKKL1
Gene Accession No. Q9UK85
Gene Alias AI414495; Cancer/testis antigen 34; CT34; dickkopf like acrosomal protein 1; dickkopf-like 1; dickkopf-like 1 (soggy); dickkopf-like protein 1; Dkkl1; mSgy; Protein soggy-1; RGD1563523; SGY; SGY1; SGY-1; Soggy; soggy 1; soggy-1; testicular secretory protein Li 15; UNQ735/PRO1429
Gene Symbols DKKL1
Host Species Rabbit
Immunogen Synthetic peptide directed towards the C-terminal of human DKKL1 (aa 182-231).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 27120
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.