Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DL-1,4-Dichloro-2,3-butanediol 98.0+%, TCI America™
SDP

Catalog No. D342325G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D342325G 25 g
D34235G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D342325G Supplier TCI America Supplier No. D342325G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 2419-73-0
Molecular Formula C4H8Cl2O2
Molecular Weight (g/mol) 159.006
InChI Key SAUBRJOIKMVSRU-UHFFFAOYSA-N
PubChem CID 102809
IUPAC Name 1,4-dichlorobutane-2,3-diol
SMILES C(C(C(CCl)O)O)Cl

Specifications

Melting Point 77°C
Color White-Yellow
Boiling Point 152°C
Quantity 25 g
Formula Weight 159.01
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material DL-1,4-Dichloro-2,3-butanediol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.