Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
4-Methylcyclohexanol, 99%, mixture of cis and trans
Click to view available options
Quantity:
250 g
1 kg
Packaging:
Glass bottle
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications
Specifications
CAS Min % | 98.5 |
CAS Max % | 100.0 |
Melting Point | -41.0°C |
Color | Colorless to Yellow |
Density | 0.9200g/mL |
Boiling Point | 171.0°C to 173.0°C |
Flash Point | 68°C |
Infrared Spectrum | Authentic |
Assay Percent Range | 99% |
Packaging | Glass bottle |
Show More |
Safety and Handling
missing translation for 'healthHazard1' | GHS Signal Word: Warning |
missing translation for 'healthHazard2' | GHS H Statement Harmful if swallowed. Harmful if inhaled. |
missing translation for 'healthHazard3' | GHS P Statement IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing. Call a POISON CENTER or doctor/physician if you feel unwell. Wear protective gloves/protective clothing/eye protection/face protect |
EINECSNumber | 209-664-8 |
TSCA | TSCA |
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction