Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

dl-1-Amino-2-propanol ≥99%, ACROS Organics™

Click to view available options
Quantity:
1kg
2.5kg
Packaging:
Glass bottle
78-96-6
C3H9NO
75.11
MFCD00008139
HXKKHQJGJAFBHI-UHFFFAOYSA-N
1-amino-2-propanol,isopropanolamine,monoisopropanolamine,threamine,2-propanol, 1-amino,2-hydroxypropylamine,amino-2-propanol,mipa,1-methyl-2-aminoethanol,2-hydroxypropanamine
4
CHEBI:19030
1-aminopropan-2-ol
CC(CN)O
78-96-6
C3H9NO
75.11
MFCD00008139
HXKKHQJGJAFBHI-UHFFFAOYSA-N
1-amino-2-propanol,isopropanolamine,monoisopropanolamine,threamine,2-propanol, 1-amino,2-hydroxypropylamine,amino-2-propanol,mipa,1-methyl-2-aminoethanol,2-hydroxypropanamine
4
CHEBI:19030
1-aminopropan-2-ol
CC(CN)O
Melting Point -2°C
Boiling Point 161°C
Density 0.9600g/mL
Specific Gravity 0.96
Flash Point 71°C
Viscosity 31 mPa.s (20°C)
Refractive Index 1.4470 to 1.449
Water 0.5% max. (K.F.)
Quantity 1kg
Infrared Spectrum Authentic
Color Undesignated
Packaging Glass bottle
Assay Percent Range 94%
Linear Formula CH3CH(OH)CH2NH2
Beilstein 04, 289
Solubility Information Solubility in water: freely soluble
Formula Weight 75.11
Physical Form Liquid
Percent Purity ≥93%
Chemical Name or Material DL-1-Amino-2-propanol, 99+%
Show More Show Less

GHS H StatementCauses severe skin burns and eye damage.Harmful in contact with skin.

GHS P StatementIF SWALLOWED: Rinse mouth.Do NOT induce vomiting.Wear protective gloves/protective clothing/eye protection/face protection.IF IN EYES: Rinse cautiously with water for several minutes.Remove contact lenses,if present and easy to do.Continue rinsing.Immediately call a POISON CENTER or doctor/physician.IF ON SKIN (or hair): Take off immediately all contaminated clothing.Rinse skin with water/shower.

;GHS P StatementIF SWALLOWED: Rinse mouth. Do NOT induce vomiting.Wear protective gloves/protective clothing/eye protection/face protection.IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses,if present and eas

EINECSNumber : 201-162-7

RTECSNumber : UA5775000

TSCA : TSCA