Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DL-10-Camphorsulfonic acid, 98%

Catalog No. AC189070025
Click to view available options
Quantity:
100 g
500 g
2.5 kg
Packaging:
Plastic bottle

CAS: 5872-08-2 | C10H16O4S | 232.29 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5872-08-2
Molecular Formula C10H16O4S
Molecular Weight (g/mol) 232.29
MDL Number MFCD00074827
InChI Key MIOPJNTWMNEORI-UHFFFAOYNA-N
Synonym reychler's acid, camphorsulfonic acid, d-camphorsulfonic acid, camphersulfosaeure, --10-camphorsulfonic acid, 2-oxobornane-10-sulphonic acid, d-10-camphorsulfonic acid, 7,7-dimethyl-2-oxobicyclo 2.2.1 heptan-1-yl methanesulfonic acid, l-camphor-10-sulfonic acid, +-10-camphorsulfonic acid
PubChem CID 18462
ChEBI CHEBI:55379
IUPAC Name (7,7-dimethyl-3-oxo-4-bicyclo[2.2.1]heptanyl)methanesulfonic acid
SMILES CC1(C2CCC1(C(=O)C2)CS(=O)(=O)O)C

Specifications

Absorbance (20% aq. soln.)
Melting Point >195°C
Color Beige
Loss on Drying 1.5% max. (105°C)
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Plastic bottle
Quantity 2.5 kg
Beilstein 11, 316
Fieser 14,71; 15,68; 16,58
Solubility Information Solubility in water: soluble. Other solubilities: slightly soluble in glacial acetic acid,slightly soluble in ethyl acetate,practically insoluble in ether
Formula Weight 232.29
Clarity (5% in water) Clear solution
Percent Purity 98%
Physical Form Granular Powder
Chemical Name or Material DL-10-Camphorsulfonic acid, 98%
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Causes severe skin burns and eye damage.
Precautionary Statement GHS P Statement
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if pre
EINECSNumber 227-527-0
TSCA TSCA

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.