Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DL-10-Camphorsulfonic acid, sodium salt, 98%, ACROS Organics™

Click to view available options
Quantity:
5g
500g
Packaging:
Glass bottle
Plastic bottle
34850-66-3
C10H15NaO4S
254.28
MFCD00135623
AWMAOFAHBPCBHJ-UHFFFAOYSA-M
sodium camphorsulfonate,sodium 7,7-dimethyl-2-oxobicyclo 2.2.1 heptan-1-yl methanesulfonate,sodium +/--10-camphorsulfonate,d-camphor-10-sulfonic acid sodium salt,sodium camphorsulfonate anion,bicyclo 2.2.1 heptane-1-methanesulfonic acid, 7,7-dimethyl-2-oxo-, sodium salt,+--sodium camphorsulfonate,sodium 2-oxobornane-10-sulphonate,sodium di-10-camphorsulfonate
23686666
sodium;(7,7-dimethyl-3-oxo-4-bicyclo[2.2.1]heptanyl)methanesulfonate
CC1(C2CCC1(C(=O)C2)CS(=O)(=O)[O-])C.[Na+]
34850-66-3
C10H15NaO4S
254.28
MFCD00135623
AWMAOFAHBPCBHJ-UHFFFAOYSA-M
sodium camphorsulfonate,sodium 7,7-dimethyl-2-oxobicyclo 2.2.1 heptan-1-yl methanesulfonate,sodium +/--10-camphorsulfonate,d-camphor-10-sulfonic acid sodium salt,sodium camphorsulfonate anion,bicyclo 2.2.1 heptane-1-methanesulfonic acid, 7,7-dimethyl-2-oxo-, sodium salt,+--sodium camphorsulfonate,sodium 2-oxobornane-10-sulphonate,sodium di-10-camphorsulfonate
23686666
sodium;(7,7-dimethyl-3-oxo-4-bicyclo[2.2.1]heptanyl)methanesulfonate
CC1(C2CCC1(C(=O)C2)CS(=O)(=O)[O-])C.[Na+]
Quantity 5g
Packaging Glass bottle
Color White
Infrared Spectrum Authentic
Melting Point 286°C to 288°C
Assay Percent Range 98%
Solubility Information Solubility in water: soluble
Formula Weight 254.28
Physical Form Amorphous Powder
Percent Purity 98%
Chemical Name or Material DL-10-Camphorsulfonic acid, sodium salt, 98%
Show More Show Less

EINECSNumber : 252-250-7

TSCA : TSCA