Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ DL 175
GREENER_CHOICE

Catalog No. 708250
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
70-825-0 50 mg
70-821-0 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 70-825-0 Supplier Tocris Bioscience™ Supplier No. 7082/50
Add to Cart
Add to Cart
This item is not returnable. View return policy

Potent and selective GPR84 agonist

DL 175 is a potent and selective GPR84 biased agonist (EC50 = 33 nM). Exhibits no significant activity in a panel of 168 other GPCRs. Exhibits bias for G protein signaling pathways. Induces morphological changes in primary murine bone marrow-derived macrophages (BMDMs) in a cellular impedance assay, and promotes phagocytosis by M1 polarized U937 cells. Induces migration of primary human monocytes, but has no effect on macrophage chemotaxis.

Specifications

Chemical Name or Material 3-(2-((4-Chloronaphthalen-1-yl)oxy)ethyl)pyridine 1-oxide
Quantity 50 mg
Target Free Fatty Acid Receptor Agonists
Molecular Formula C17H14ClNO2
Formula Weight 4495
Purity 98%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.