Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DL-2,3-Diaminopropionic Acid Hydrobromide 98.0+%, TCI America™
SDP

Catalog No. D011525G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D011525G 25 g
D01155G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D011525G Supplier TCI America Supplier No. D011525G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 18635-45-5
Molecular Formula C3H9BrN2O2
Molecular Weight (g/mol) 185.02
MDL Number MFCD00012883
InChI Key IZNPALADKWAZMW-UHFFFAOYNA-N
Synonym 2,3-diaminopropanoic acid hydrobromide, 2,3-diaminopropionic acid hydrobromide, dl-2,3-diaminopropionic acid hydrobromide, 2,3-diaminopropionic acid hbr, 2,3-diaminopropanoic acid hbr, 2,3-diaminopropionic acid monohydrobromide, 3-amino-dl-alanine hydrobromide, 2,3-diaminopropanoic acid, bromide, 2,3-diaminopropionic acid hydro-bromide, dl-2,3-diaminopropionicacidhydrobromide
PubChem CID 3084297
IUPAC Name hydrogen 2,3-diaminopropanoic acid bromide
SMILES [H+].[Br-].NCC(N)C(O)=O

Specifications

Melting Point 230°C
Color White-Yellow
Quantity 25 g
Formula Weight 185.02
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material DL-2,3-Diaminopropionic Acid Hydrobromide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.