Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DL-2,3-Diaminopropionic acid monohydrochloride, 99%

Catalog No. AC204670050
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle

CAS: 54897-59-5 | C3H8N2O2·HCl | 140.57 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 54897-59-5
Molecular Formula C3H8N2O2·HCl
Molecular Weight (g/mol) 140.57
MDL Number MFCD00012884
InChI Key SKWCZPYWFRTSDD-UHFFFAOYSA-N
Synonym 2,3-diaminopropanoic acid hydrochloride, dl-2,3-diaminopropionic acid hydrochloride, 3-amino-dl-alanine hydrochloride, dl-2,3-diaminopropionic acid monohydrochloride, 2,3-diaminopropionic acid hydrochloride, +--2,3-diaminopropionic acid hydrochloride, 3-amino-dl-alanine monohydrochloride, 2,3-diaminopropionic acid monohydrochloride, dl-2,3-diaminopropanoic acid hydrochloride, diaminopropionic acid
PubChem CID 108638
IUPAC Name 2,3-diaminopropanoic acid;hydrochloride
SMILES C(C(C(=O)O)N)N.Cl

Specifications

Melting Point 222°C to 226°C
Color White to Beige
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (Argentometry)
Packaging Glass bottle
Linear Formula H2NCH2CH(NH2)CO2H·HCl
Quantity 5 g
Beilstein 04, 405
Merck Index 10, 2948
Solubility Information Solubility in water: freely soluble in water
Formula Weight 140.57
Percent Purity 99%
Physical Form Fine Crystalline Powder
Chemical Name or Material DL-2, 3-Diaminopropionic acid monohydrochloride
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 259-387-1

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.